Learn More
Cayman Chemical TMPyP4 (tosylate), Purity: ≥95%, CAS: 36951-72-1, 50 mg
SDP
Catalog No. 502966384
Encompass_Preferred
A cationic porphyrin that intercalates into and stabilizes G-quadruplexes; inhibits telomerase activity (IC50 ≤ 50 µM), blocking the clonogenic growth and migration of cancer cells in culture and in vivo
Catalog No. 50-296-6384 Supplier Cayman Chemical Supplier No. 1847450 MG
A cationic porphyrin that intercalates into and stabilizes G-quadruplexes; inhibits telomerase activity (IC50 ≤ 50 µM), blocking the clonogenic growth and migration of cancer cells in culture and in vivo