Learn More
Biotium 5-(and-6-)-((N-(5-aminopentyl)amino)carbonyl) Tetramethylrhodamine (Tetramethylrhodamine cadaverine) (mixed isomer)
Catalog No. 501964836
Encompass_Preferred
INA (5-iodonaphthyl-1-azide) is a lipophilic photoreactive probe, which has been used to selectively label membrane-embedded cysteine residues of proteins. Properties: Light yellow solid soluble in DMSO or DMF; Store at 4°C and protect from light; C30H34N4O4; MW: 295.08.
Catalog No. 50-196-4836 Supplier Biotium Supplier No. 92001
INA (5-iodonaphthyl-1-azide) is a lipophilic photoreactive probe, which has been used to selectively label membrane-embedded cysteine residues of proteins. Properties: Light yellow solid soluble in DMSO or DMF; Store at 4°C and protect from light; C30H34N4O4; MW: 295.08.