Learn More
Biotium TMRE, 2 mM solution in DMSO
Catalog No. 501964741
Encompass_Preferred
TMRE (Tetramethylrhodamine ethyl ester, perchlorate) and TMRM are preferred dyes for quantitative measurements of membrane potentials using the Nernst equation. TMRE has been used to separate viable and non-apoptotic cells from cell sorting samples. TMRE staining is reversible and does not affect cell proliferation and viability. TMRE is also available in solid form.Properties: Ex/Em (MeOH) = 549/574 nm; Store the solution at 4°C and protect from light; C26H27ClIN2O7; MWt: 515; [115532-52-0];
Catalog No. 50-196-4741 Supplier Biotium Supplier No. 70005
TMRE (Tetramethylrhodamine ethyl ester, perchlorate) and TMRM are preferred dyes for quantitative measurements of membrane potentials using the Nernst equation. TMRE has been used to separate viable and non-apoptotic cells from cell sorting samples. TMRE staining is reversible and does not affect cell proliferation and viability. TMRE is also available in solid form.Properties: Ex/Em (MeOH) = 549/574 nm; Store the solution at 4°C and protect from light; C26H27ClIN2O7; MWt: 515; [115532-52-0];