Learn More
Biotium Tetramethylrhodamine Ethyl Ester, Perchlorate
Catalog No. 501964749
Encompass_Preferred
TMRE (Tetramethylrhodamine ethyl ester, perchlorate) and TMRM are preferred dyes for quantitative measurements of membrane potentials using the Nernst equation. The dyes do not form aggregates in cell membranes and have minimal interaction with membrane proteins. Thus, the transmembrane distribution of the dyes is directly related to the membrane potential according to the Nernst equation. Biotium also offers TMRE, 2 mM in DMSO as a ready to use stock solution.; ; Ex/Em (MeOH) = 549/574 nm; Red solid soluble in DMSO, DMF or EtOH; Store at 4°C and protect from light; C26H27ClN2O7; MW: 515; [115532-52-0];
Catalog No. 50-196-4749 Supplier Biotium Supplier No. 70016
TMRE (Tetramethylrhodamine ethyl ester, perchlorate) and TMRM are preferred dyes for quantitative measurements of membrane potentials using the Nernst equation. The dyes do not form aggregates in cell membranes and have minimal interaction with membrane proteins. Thus, the transmembrane distribution of the dyes is directly related to the membrane potential according to the Nernst equation. Biotium also offers TMRE, 2 mM in DMSO as a ready to use stock solution.; ; Ex/Em (MeOH) = 549/574 nm; Red solid soluble in DMSO, DMF or EtOH; Store at 4°C and protect from light; C26H27ClN2O7; MW: 515; [115532-52-0];