Learn More
Medchemexpress LLC HY-D0985A 25mg , TMRE CAS:115532-52-0 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HYD0985A25MG
Encompass_Preferred
HY-D0985A 25mg TMRE CAS: 115532-52-0 TMRE is a mitochondria specific dye (λex=550 nm, λem=575 nm). Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-226-1161