Learn More
Medchemexpress LLC HY-D0985A 5mg Medchemexpress, TMRE CAS:115532-52-0 Purity:>98%
SDP
Catalog No. 501933227
Encompass_Preferred
Medchemexpress, HY-D0985A 5mg TMRE CAS:115532-52-0 TMRE is a mitochondria specific dye (λ ex =550 nm, λ em =575 nm). Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-193-3227 Supplier Medchemexpress LLC Supplier No. HYD0985A5MG
Medchemexpress, HY-D0985A 5mg TMRE CAS:115532-52-0 TMRE is a mitochondria specific dye (λ ex =550 nm, λ em =575 nm). Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.