Learn More
Abcam Tetramethylrhodamine, methyl ester (TMRM), 10MG
Supplier:  Abcam AB27554710MG
Encompass_Preferred
MW 436.93 g/mol, Purity >95%. Fluorescent, cell-permeant cationic dye for measuring mitochondrial membrane potential. Accumulates in healthy, negatively charged mitochondria. Exhibits re-orange fluorscence (Em = 573 nm). Minimal fluorescence observed when mitochondrial membrane potential collapses. Used to monitor cell health and viability. Stained cells can be analysed via flow cytometry, fluorescence microscopy or fluorescence plate reader techniques. .
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. 50-113-9332