Learn More
Biotium Tetramethylrhodamine Methyl Ester, Perchlorate
Supplier:  Biotium 70017
Encompass_Preferred
TMRM (Tetramethylrhodamine methyl ester, perchlorate) and TMRE are preferred dyes for quantitative measurements of membrane potentials using the Nernst equation. The dyes do not form aggregates in cell membranes and have minimal interaction with membrane proteins. Thus, the transmembrane distribution of the dyes is directly related to the membrane potential according to the Nernst equation.Properties: Ex/Em (MeOH) = 548/573 nm; Red solid soluble in DMSO, DMF or EtOH; Store at 4°C and protect from light; C25H25ClN2O7; MW: 501; [115532-50-8].
Catalog No. 50-196-4750