Learn More
Medchemexpress LLC TMRM Perchlorate | 115532-50-8 | 99.7% | 1 ML
SDP
Catalog No. 5000410607
Encompass_Preferred
TMRM Perchlorate is a rapidly and selectively mitochondrial membrane potential fluorescent probe that exhibits red fluorescence. Rhodamine dyes, including TMRM Perchlorate, are membrane-permeable cationic fluorescent probes that specifically recognize mitochondrial membrane potentials, attaching to mitochondria and producing bright fluorescence. At certain concentrations, rhodamine dyes demonstrate low toxicity to cells, making them suitable for detecting mitochondria in animal cells, plant cells, and microorganisms.
  • Membrane-permeable cationic fluorescent probe.
  • Specifically recognizes mitochondrial membrane potentials.
  • Attaches to mitochondria and produces bright fluorescence.
  • Low toxicity to cells at certain concentrations.
  • Used to detect mitochondria in animal cells, plant cells, and microorganisms.
  • Rapidly and selectively targets mitochondrial membrane potential.
Catalog No. 50-004-10607 Supplier Medchemexpress LLC Supplier No. HYD0984A10MM/1ML
Add to Cart
Add to Cart
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
TMRM Perchlorate is a rapidly and selectively mitochondrial membrane potential fluorescent probe that exhibits red fluorescence. Rhodamine dyes, including TMRM Perchlorate, are membrane-permeable cationic fluorescent probes that specifically recognize mitochondrial membrane potentials, attaching to mitochondria and producing bright fluorescence. At certain concentrations, rhodamine dyes demonstrate low toxicity to cells, making them suitable for detecting mitochondria in animal cells, plant cells, and microorganisms.
  • Membrane-permeable cationic fluorescent probe.
  • Specifically recognizes mitochondrial membrane potentials.
  • Attaches to mitochondria and produces bright fluorescence.
  • Low toxicity to cells at certain concentrations.
  • Used to detect mitochondria in animal cells, plant cells, and microorganisms.
  • Rapidly and selectively targets mitochondrial membrane potential.