Learn More
Medchemexpress LLC TMRM Perchlorate | 115532-50-8 | 99.7% | 100 MG
SDP
Catalog No. 5000409614
Encompass_Preferred
TMRM Perchlorate is a rhodamine dye, a membrane-permeable cationic fluorescent probe that specifically recognizes mitochondrial membrane potentials. It attaches to mitochondria, producing bright fluorescence, and is commonly used to detect mitochondria in animal cells, plant cells, and microorganisms. It is described as a rapidly and selectively mitochondrial membrane potential fluorescent probe with red fluorescence.
  • Fluorescent dye
  • Rapidly and selectively targets mitochondrial membrane potential
  • Produces red fluorescence
  • Low toxicity to cells at certain concentrations
Catalog No. 50-004-09614 Supplier Medchemexpress LLC Supplier No. HYD0984A100MG
TMRM Perchlorate is a rhodamine dye, a membrane-permeable cationic fluorescent probe that specifically recognizes mitochondrial membrane potentials. It attaches to mitochondria, producing bright fluorescence, and is commonly used to detect mitochondria in animal cells, plant cells, and microorganisms. It is described as a rapidly and selectively mitochondrial membrane potential fluorescent probe with red fluorescence.
  • Fluorescent dye
  • Rapidly and selectively targets mitochondrial membrane potential
  • Produces red fluorescence
  • Low toxicity to cells at certain concentrations