Learn More
TARGETMOL CHEMICALS INC TMRM PERCHLORATE 25MG
SDP
Catalog No. 502577318
Encompass_Preferred
Also available in 5 mg 10 mg 50 mg 100 mg 200 mg 500 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. TMRM Perchlorate (T668) is a cell-permeant cationic lipophilic red fluorescent dye ((lambda)ex=530 nm (lambda)em=592 nm). purity: 99%
Catalog No. 50-257-7318 Supplier TARGETMOL CHEMICALS INC Supplier No. T1710925MG
Also available in 5 mg 10 mg 50 mg 100 mg 200 mg 500 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. TMRM Perchlorate (T668) is a cell-permeant cationic lipophilic red fluorescent dye ((lambda)ex=530 nm (lambda)em=592 nm). purity: 99%