Learn More
Medchemexpress LLC HY-D0984A 5mg Medchemexpress, TMRM Perchlorate CAS:115532-50-8 Purity:>98%
SDP
Catalog No. 501933226
Encompass_Preferred
Medchemexpress, HY-D0984A 5mg TMRM Perchlorate CAS:115532-50-8 TMRM Perchlorate is a cell-permeant cationic lipophilic red fluorescent dye (λ ex =530 nm, λ em =592 nm). Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-193-3226 Supplier Medchemexpress LLC Supplier No. HYD0984A5MG
Medchemexpress, HY-D0984A 5mg TMRM Perchlorate CAS:115532-50-8 TMRM Perchlorate is a cell-permeant cationic lipophilic red fluorescent dye (λ ex =530 nm, λ em =592 nm). Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.