Learn More
Aat Bioquest TMRM [Tetramethylrhodamine methyl ester] CAS 115532-50-8 (25 mg)
SDP
Catalog No. 502250058
Encompass_Preferred
Positively charged rhodamine dyes such as rhodamine esters and rosamines are selectively localized in mitochondria thus they are widely used for labeling mitochondria of live cells. This catalog is for 25 mg.
This item is not returnable. View return policy
Catalog No. 50-225-0058 Supplier Aat Bioquest Supplier No. 22221
Positively charged rhodamine dyes such as rhodamine esters and rosamines are selectively localized in mitochondria thus they are widely used for labeling mitochondria of live cells. This catalog is for 25 mg.