Learn More
YMC America, Inc TMS 120A 3um 100x0.3mm 1/32
SDP
Catalog No. 5000879750
Encompass
YMC-Pack TMS columns are bonded with trimethylmonochlorosilane to create a phase with intermediate polarity This makes for a versatile stationary phase that is useful for analysis of extremely hydrophobic compounds using conventional reversed-phase solvents or highly polar compounds using high aqueous solvents The unique chemistry of this packing material is also well-suited for the analysis of multifunctional compounds Selectivity characteristics of a C1 bonded phase can be unique and samples must be evaluated to determine the applicability of the phase
Catalog No. 50-008-79750 Supplier YMC America, Inc Supplier No. TM12S0310H0RU
YMC-Pack TMS columns are bonded with trimethylmonochlorosilane to create a phase with intermediate polarity This makes for a versatile stationary phase that is useful for analysis of extremely hydrophobic compounds using conventional reversed-phase solvents or highly polar compounds using high aqueous solvents The unique chemistry of this packing material is also well-suited for the analysis of multifunctional compounds Selectivity characteristics of a C1 bonded phase can be unique and samples must be evaluated to determine the applicability of the phase