Learn More
TARGETMOL CHEMICALS INC TMS 25MG
SDP
Catalog No. 502580208
Encompass_Preferred
Also available in 5 mg 10 mg 50 mg 100 mg 500 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. TMS (23'45'-Tetramethoxystilbene) is a very selective and potent competitive inhibitor of P450 1B1 (CYP1A1). purity: 99%
Catalog No. 50-258-0208 Supplier TARGETMOL CHEMICALS INC Supplier No. T358525MG
Also available in 5 mg 10 mg 50 mg 100 mg 500 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. TMS (23'45'-Tetramethoxystilbene) is a very selective and potent competitive inhibitor of P450 1B1 (CYP1A1). purity: 99%