Learn More
Medchemexpress LLC HY-19340 5mg Medchemexpress, TMS CAS:24144-92-1 Purity:>98%
SDP
Catalog No. 502082119
Encompass_Preferred
Medchemexpress, HY-19340 5mg TMS CAS:24144-92-1 TMS ((E)-2,3',4,5'-tetramethoxystilbene) is a selective and competitive CYP1B1 inhibitor with an IC50 of 6 nM and a Ki value of 3 nM. TMS shows a lesser extent inhibitory effect on CYP1A1 (IC5098% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-208-2119 Supplier Medchemexpress LLC Supplier No. HY193405MG
Medchemexpress, HY-19340 5mg TMS CAS:24144-92-1 TMS ((E)-2,3',4,5'-tetramethoxystilbene) is a selective and competitive CYP1B1 inhibitor with an IC50 of 6 nM and a Ki value of 3 nM. TMS shows a lesser extent inhibitory effect on CYP1A1 (IC5098% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.