Learn More
Enzo Life Sciences Prothymosin alpha (human) monoclonal antibody (1G8) (100µg)
SDP
Supplier:  Enzo Life Sciences ALX8049160100
Encompass_Preferred
Clone: 1G8. Isotype: IgG2a. Immunogen: Recombinant human prothymosin α (aa 1-110) with GST tag. Species reactivity: Human. Applications: ELISA, WB. Purity: Protein A-affinity purified. Formulation: Liquid. In PBS, pH 7.2.
Catalog No. 50-201-0024