Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC TMSB4X/Thymosin beta 50ug
SDP

Catalog No. 5000195669
Encompass_Preferred

Recombinant human Thymosin beta-4 (TMSB4X) is a small actin-binding peptide supplied as a His-tagged recombinant protein for research use. The product includes sequence and molecular weight information, recommended reconstitution and storage guidance, and analytical purity data appropriate for biochemical and cell biology assays.

  • Acts as an actin-binding peptide that modulates actin dynamics.
  • Recombinant human sequence, approximately 12 kDa.
  • His tag to facilitate purification and detection.
  • Greater than 95% purity by reducing SDS-PAGE.
  • Expressed in Escherichia coli for efficient production.
  • Stability and handling recommendations include freezing aliquots at -20°C or -80°C.
  • Reconstitute to 100 μg/mL per datasheet instructions.

Catalog No. 50-001-95669 Supplier Medchemexpress LLC Supplier No. HYP71930A50UG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

Recombinant human Thymosin beta-4 (TMSB4X) is a small actin-binding peptide supplied as a His-tagged recombinant protein for research use. The product includes sequence and molecular weight information, recommended reconstitution and storage guidance, and analytical purity data appropriate for biochemical and cell biology assays.

  • Acts as an actin-binding peptide that modulates actin dynamics.
  • Recombinant human sequence, approximately 12 kDa.
  • His tag to facilitate purification and detection.
  • Greater than 95% purity by reducing SDS-PAGE.
  • Expressed in Escherichia coli for efficient production.
  • Stability and handling recommendations include freezing aliquots at -20°C or -80°C.
  • Reconstitute to 100 μg/mL per datasheet instructions.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.