Learn More
Medchemexpress LLC TMSB4X/Thymosin beta 5ug
SDP
Catalog No. 5000195740
Encompass_Preferred
Recombinant human Thymosin beta 4 (TMSB4X) is a small, His-tagged protein expressed in E. coli and supplied as a lyophilized powder for research applications. The protein binds G-actin to regulate actin dynamics and is commonly used in studies of cytoskeletal organization, cell migration, proliferation, and differentiation. Formulation and storage guidance are provided for optimal activity and low endotoxin performance.
  • Acts as an actin-binding regulator affecting cytoskeletal organization.
  • Supplied as a lyophilized, His-tagged recombinant human protein.
  • Expressed in E. coli with sequence matching the human peptide.
  • Low endotoxin level (<1 EU/μg) suitable for sensitive assays.
  • Lyophilized from Tris-HCl/NaCl buffer; reconstitute at ≥100 μg/mL and add carrier for long-term storage.
  • Stable at -20°C; aliquot and avoid repeated freeze-thaw cycles.
Catalog No. 50-001-95740 Supplier Medchemexpress LLC Supplier No. HYP71930A5UG
Recombinant human Thymosin beta 4 (TMSB4X) is a small, His-tagged protein expressed in E. coli and supplied as a lyophilized powder for research applications. The protein binds G-actin to regulate actin dynamics and is commonly used in studies of cytoskeletal organization, cell migration, proliferation, and differentiation. Formulation and storage guidance are provided for optimal activity and low endotoxin performance.
  • Acts as an actin-binding regulator affecting cytoskeletal organization.
  • Supplied as a lyophilized, His-tagged recombinant human protein.
  • Expressed in E. coli with sequence matching the human peptide.
  • Low endotoxin level (<1 EU/μg) suitable for sensitive assays.
  • Lyophilized from Tris-HCl/NaCl buffer; reconstitute at ≥100 μg/mL and add carrier for long-term storage.
  • Stable at -20°C; aliquot and avoid repeated freeze-thaw cycles.