Learn More
Medchemexpress LLC TMSB4X/Thymosin beta 5ug
SDP
Supplier:  Medchemexpress LLC HYP727765UG
Encompass_Preferred
TMSB4X proteins play a critical role in cytoskeletal organization influencing actin dynamics by binding and sequestering actin monomers As a potent inhibitor of actin polymerization it affects the cytoskeleton TMSB4X/Thymosin beta 4 Protein Human is the recombinant human-derived Thymosin beta 4 protein expressed by E coli with tag free
Catalog No. 50-001-81760