Learn More
Aviva Systems Biology Corp TMTC1 Antibody - middle region (ARP49127_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP49127_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against TMTC1. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the middle region of human TMTC1. Alternate names: ARG99, FLJ31400, FLJ41625, OLF, TMTC1AApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat, Yeast
Catalog No. 50-014-60025