Learn More
Aviva Systems Biology Corp TMTC4 Antibody - C-terminal region (ARP45028_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP45028_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against TMTC4. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the C terminal region of human TMTC4. Alternate names: FLJ14624, FLJ22153ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat, Zebrafish
Catalog No. 50-014-53443