Learn More
Thermo Scientific™ TMTpro 18-plex, TMTpro-134C, and TMTpro-135N Label Reagents
Shop All Thermo Scientific ProductsDescription
TMTpro label reagents are the next generation of tandem mass tags, designed to increase the level of sample multiplexing without compromising on protein identification and quantitation. The Thermo Scientific TMTpro 18plex Label Reagent Set enables multiplexing up to 18 samples, enabling even greater throughput for protein identification and quantitative analysis by tandem mass spectrometry (MS).
The structures of the TMTpro and TMT tags are similar in being isobaric and amine reactive, but the TMTpro tag has a longer spacer region and isobutyl proline mass reporter. After MS/MS fragmentation, each TMTpro 18plex tag generates a unique reporter mass (e.g., TMTpro 126-135 Da) in the low-mass region of the high-resolution MS/MS spectrum that is used for relative quantitation of protein expression levels.
The TMT pro134C and TMTpro135N Label Reagents are identical in composition and structure to the other TMTpro 16plex tags, but are not isobaric. Addition of the TMTpro 134C and 135N tags create a 17th and 18th channel for relative quantitation using high resolution Orbitrap MS instruments. These tags are available separately (Cat. No. A52046, A52048), as part of the TMTpro 18plex Label Reagent Set, or as part of the complete 18plex set of reagents in a 96-well plate format.
Features of the TMTpro 18plex Label Reagent Set include:
- Multiplex—concurrent MS analysis of up to 18 samples derived from cells, tissues, or biological fluids
- Robust—increased multiplex capability results in fewer missing quantitative values among samples and higher confidence among replicates
- Efficient—amine-reactive, NHS ester-activated reagents ensure greater than 95% labeling of all peptides regardless of protein sequence or proteolytic enzyme specificity
- Optimized for use—with high resolution Thermo Scientific MS/MS Orbitrap platforms, analysis fully supported by Proteome Discoverer 2.3
- Convenient size—provided in a ready-to-use, single-use format to ensure optimal stability and performance
For high resolution analysis of TMTpro-labeled peptides, the recommended LC column for the Nanospray Flex source is the Acclaim PepMap 100 C18 LC Column (Cat. No. 164942 or 164939). For the EASY-Spray source, the recommended LC column is the EASY-Spray C18 LC Column (Cat. No. ES902 or ES903). When combined with the industry-leading, high resolution Orbitrap instruments and software, TMTpro reagents provide an integrated total solution for quantitative protein expression analysis.
Specifications
Specifications
| Formulation | Liquid in stabilized DMSO solution |
| For Use With (Equipment) | Mass Spectrometer |
| Product Type | Isobaric Label Reagent Set |
| Product Line | TMT |
| Sufficient For | 72 Reactions |
| Content And Storage | Sufficient for: Four 18-plex (4 x 18-way) experiments |
| Label or Dye | TMTpro 18-plex |
| Starting Material | Cell Lysate |
| Final Product Type | Protein PEPTIDE |
| Detection Method | Mass Spectrometry |
| Show More |
Frequently Asked Questions (FAQs)
TMT and TMTpro labels are not compatible with buffers containing primary amines. After protein digestion, peptides need to be cleaned up and dried down. Prior to labeling, we do recommend resuspending the peptides in 10 µL of either 100 mM TEAB (pH 8.5) or 100 mM HEPES (pH 8.0).
For further steps after TMT labelling, the stabilizing solution can be considered as a 33% DMSO solution. If subsequent excess TMT removal will be performed using EasyPep solid-phase extraction (SPE), the DMSO solution does not need to be diluted. On the other hand, the use of any reverse phase chromatography method (i.e., Cat. No. 84868) might need dilution of the sample to ensure that the DMSO concentration is tolerated by the resin. Alternatively, the peptide mix can be dried down using Speed Vac and then resuspended in the corresponding buffer for a particular column (Reverse Phase or Desalting).
TMT and TMTpro label reagents in 96-well plates are optimized to work with 1 to 10 µg of protein digest.
Yes, the sealing foil can be partially peeled to access individual sets of TMT or TMTpro labels. Alternatively, each of the wells can be perforated to transfer the TMT/TMTpro labels to a different tube, if needed.
All the plates are barcoded in the long lower edge, helping to recognize correct plate orientation, plus the plate has a notch on the left corners.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.