Learn More
Medchemexpress LLC TMX-2039 | 2488892-01-7 | C17H20BrFN6O3S | 50 MG
SDP
Catalog No. 5000375833
Encompass_Preferred
TMX-2039 is a pan-CDK inhibitor targeting both cell cycle CDKs (CDK1, CDK2, CDK4, CDK5, and CDK6) and transcriptional CDKs (CDK7 and CDK9). It exhibits high potency with low nanomolar IC50 values against several CDKs and can also function as a ligand for target proteins in PROTAC applications.
  • Broad-spectrum CDK inhibition: Inhibits multiple cyclin-dependent kinases involved in cell cycle progression and gene transcription.
  • High potency: Demonstrates low nanomolar IC50s against several CDKs, particularly CDK2 and CDK5.
  • PROTAC ligand: Can be used as a ligand for target proteins in PROTAC (Proteolysis Targeting Chimeras) applications.
  • High purity: Reported purity of 99.42%.
  • Research use only: Suitable for research applications.
Catalog No. 50-003-75833 Supplier Medchemexpress LLC Supplier No. HY15106950MG
TMX-2039 is a pan-CDK inhibitor targeting both cell cycle CDKs (CDK1, CDK2, CDK4, CDK5, and CDK6) and transcriptional CDKs (CDK7 and CDK9). It exhibits high potency with low nanomolar IC50 values against several CDKs and can also function as a ligand for target proteins in PROTAC applications.
  • Broad-spectrum CDK inhibition: Inhibits multiple cyclin-dependent kinases involved in cell cycle progression and gene transcription.
  • High potency: Demonstrates low nanomolar IC50s against several CDKs, particularly CDK2 and CDK5.
  • PROTAC ligand: Can be used as a ligand for target proteins in PROTAC (Proteolysis Targeting Chimeras) applications.
  • High purity: Reported purity of 99.42%.
  • Research use only: Suitable for research applications.