Learn More
Medchemexpress LLC HY-145321 5mg , TMX-4100 CAS:2367619-63-2 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY1453215MG
Encompass_Preferred
HY-145321 5mg TMX-4100 CAS: 2367619-63-2 TMX-4100 is a selective phosphodiesterase 6D (PDE6D) degrader. TMX-4100 shows a high degradation preference for PDE6D with the DC50 values less than 200 nM in MOLT4, Jurkat, and MM.1S cells. TMX-4100 can be used for the research of multiple myeloma. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-226-1859