Learn More
Medchemexpress LLC Tmx-4116 | 2766385-56-0 | 99.8% | C17H19N5O4S | 10MG
SDP
Catalog No. 5000261195
Encompass_Preferred
TMX-4116 is a small-molecule degrader of casein kinase 1α (CK1α) intended for preclinical research. It selectively induces degradation of CK1α (DC50 <200 nM in MOLT4, Jurkat, and MM.1S cells), is supplied as a solid with analytical documentation, and includes recommended solubility and storage guidance.
Catalog No. 50-002-61195 Supplier Medchemexpress LLC Supplier No. HY14532210MG
TMX-4116 is a small-molecule degrader of casein kinase 1α (CK1α) intended for preclinical research. It selectively induces degradation of CK1α (DC50 <200 nM in MOLT4, Jurkat, and MM.1S cells), is supplied as a solid with analytical documentation, and includes recommended solubility and storage guidance.