Learn More
Aviva Systems Biology Corp TMX1 Antibody - C-terminal region (ARP62938_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP62938_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against TMX1. It was validated on Western Blot. . Alternate names: DKFZp564E1962, PDIA11, TMX, TXNDC, TXNDC1ApplicationWB. Species reactivity: Cow, Dog, Horse, Human, Pig
Catalog No. 50-287-814