Learn More
Stellar Scientific True North® Cryo Vials. 2.0mL, Standing, Colored Lids, External Thread, 500/CS
SDP
Catalog No. NC1374558
Encompass
Innovative design saves space in your freezer!Our externally-threaded vial with a flush cap design allows you to easily fill up all standard-sized cryogenic storage boxes including the 10 x 10 format. Using True North® Cryogenic Vials can increase your freezers holding capacity by up to 23% against other major brands.Superior puncture- and rupture-resistance polypropylene with silicon lid sealPatented lid with ergonomic 1.5 twist to open and closePrinted writing area and graduationsInner resealable packs of 50 vials, complete with traceable sterility certificateRecommended for mechanical freezers and vapor phase LN2 (-196°)Sterile, non-pyrogenic and non-cytotoxic2.0 mL vial measures 13 mm dia x 49 mm H, 4.0 mL 13 mm dia x 88 mm H
Catalog No. NC1374558 Supplier Stellar Scientific Supplier No. STSHS23202A
Innovative design saves space in your freezer!Our externally-threaded vial with a flush cap design allows you to easily fill up all standard-sized cryogenic storage boxes including the 10 x 10 format. Using True North® Cryogenic Vials can increase your freezers holding capacity by up to 23% against other major brands.Superior puncture- and rupture-resistance polypropylene with silicon lid sealPatented lid with ergonomic 1.5 twist to open and closePrinted writing area and graduationsInner resealable packs of 50 vials, complete with traceable sterility certificateRecommended for mechanical freezers and vapor phase LN2 (-196°)Sterile, non-pyrogenic and non-cytotoxic2.0 mL vial measures 13 mm dia x 49 mm H, 4.0 mL 13 mm dia x 88 mm H