Learn More
Seracare Life Sciences Inc Seraseq TNA (DNA/RNA) WT Mix | 2 x 20 µl
Catalog No. NC1835276
Encompass
Seracare has developed two formats of total nucleic acid (TNA) derived from GM24385 human cell line as negative sample reference materials for use in targeted NGS assays. The cell line is applied as the WT background in all existing Seraseq tumor DNA and RNA products. As a negative reference material, they have extracted DNA and RNA from GM24385 as a buffered mix (TNA WT) as well as a formalin fixed and paraffin embedded option (FFPE curls) to support labs validating assay specificity at wildtype base positions, as well as specificity at positions affected by mutations included in all of the Seraseq RNA and DNA (Mutation Mix and Reference Material) products.
This item is not returnable. View return policy
Catalog No. NC1835276 Supplier Seracare Life Sciences Inc Supplier No. 07101580
Seracare has developed two formats of total nucleic acid (TNA) derived from GM24385 human cell line as negative sample reference materials for use in targeted NGS assays. The cell line is applied as the WT background in all existing Seraseq tumor DNA and RNA products. As a negative reference material, they have extracted DNA and RNA from GM24385 as a buffered mix (TNA WT) as well as a formalin fixed and paraffin embedded option (FFPE curls) to support labs validating assay specificity at wildtype base positions, as well as specificity at positions affected by mutations included in all of the Seraseq RNA and DNA (Mutation Mix and Reference Material) products.