Learn More
Abcam TNB, Yellow chromogenic standard for DTNB thiol quantification, 250MG
Catalog No. 502204979
Encompass_Preferred
MW 199.19 Da, Purity =95%. TNB, Yellow chromogenic standard for DTNB thiol quantification. Achieve your results faster with highly validated, pure and trusted compounds.
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. 50-220-4979 Supplier Abcam Supplier No. AB145632250MG
MW 199.19 Da, Purity =95%. TNB, Yellow chromogenic standard for DTNB thiol quantification. Achieve your results faster with highly validated, pure and trusted compounds.
The product is subject to the following: Abcam Restricted Use Statement