Learn More
enQuire Bioreagents Calponin-1 Anti-Human Monoclonal [Clone SPM169], Size: 100ug
Supplier:  enQuire Bioreagents 3371MSM1P1
Encompass_Preferred
Product Type: Antibody. Application: Flow, IF, IHC. Conjugate/Tag: Unconjugated. Clone: T2H5. Clonality: Monoclonal. Host: Mouse. Isotype: IgG1 kappa. Positive Control: Extracellular matrix in tonsil, blood vessels. Stroma of many tumors such as breast, squamous cell,, lung carcinomas. Staining of normal fibroblasts serves as internal positive control. Cellular Staining: Connective tissue matrix. Buffer: 1mM PBS with 0.05% BSA & 0.05% azide. Concentration: 200ug/ml. Purity: Protein A/G Purified. Immunogen: Human breast carcinoma. Storage: Short term: Store at 2-4C. Long term, aliquot and store at -20C. MW: 210kDa, 300kDa.Chromosome: 9q33. Gene Symbol: TNC. Gene ID: 3371. Ensemble: ENSG00000041982. Uniprot: NP_002151.
Catalog No. 50-188-4789