Learn More
Teknova TNE Buffer, 1000mL, Sterile. 2 pack.
SDP
Catalog No. 502387900
Encompass
100mM Sodium Chloride, 10mM Tris, 1mM EDTA
Catalog No. 50-238-7900 Supplier Teknova Supplier No. T05602PK
100mM Sodium Chloride, 10mM Tris, 1mM EDTA