Learn More
Quality Biological Inc TNE Buffer 500mL
SDP
Catalog No. 502209780
Encompass_Preferred
TNE Buffer is often used as a base material to generate lysis buffers. Filtered through a 0.22 micron membrane DNase Rnase and Protease tested Manufactured in an ISO 90012015 certified facility.
Catalog No. 50-220-9780 Supplier Quality Biological Inc Supplier No. 351302101
TNE Buffer is often used as a base material to generate lysis buffers. Filtered through a 0.22 micron membrane DNase Rnase and Protease tested Manufactured in an ISO 90012015 certified facility.