Learn More
Enzo Life Sciences TNF-alpha (human), (recombinant) (cell culture grade) (10µg). CAS: 94948-59-1
SDP
Supplier:  Enzo Life Sciences ALX520002C010
Encompass_Preferred
Alternative name: Tumor necrosis factor alpha, TNFSF 2. Purity: ≥97% (SDS-PAGE). Formulation: Lyophilized from 0.2μm-filtered solution containing mannitol. Source: Produced in yeast. Long Term Storage: -20°C.
Catalog No. 50-200-9348