Learn More
Enzo Life Sciences TNF-alpha (human) monoclonal antibody (TNF-D) (100µg)
SDP
Supplier:  Enzo Life Sciences ALX804199C100
Encompass_Preferred
Clone: TNF-D. Isotype: IgG1. Immunogen: Recombinant human TNF-α (tumor necrosis factor-α). Species reactivity: Human. Applications: ELISA, Flow Cytometry, FUNC. Formulation: Liquid. In PBS.
Catalog No. 50-200-9822