Learn More
Enzo Life Sciences TNF-alpha Recombinant monoclonal antibody (cA2) (Rabbit IgG kappa) (200µg)
SDP
Supplier:  Enzo Life Sciences ENZABS5610200
Encompass_Preferred
Clone: cA2 (Infliximab). Isotype: IgGκ. Immunogen: Purified recombinant human TNF (rTNF). Species reactivity: Human. Specificity: Binds to soluble and transmembrane forms of TNF-α and neutralizes its biological activity. Applications: ELISA, Flow Cytometry in vivo Assay. Purity: Protein A affinity purified. Formulation: Liquid. In PBS containing 0.02% Proclin 300.
Catalog No. 50-201-3087