Learn More
Enzo Life Sciences TNF-alpha Recombinant monoclonal antibody (D2E7) (Human IgG1 kappa) (200µg)
SDP
Supplier:  Enzo Life Sciences ENZABS5660200
Encompass_Preferred
Clone: D2E7 (Adalimumab). Isotype: IgG1κ. Immunogen: This antibody was generated by affinity maturation from the 2SD4 scFv, which in turn was selected on human TNF alpha, using the murine anti-hTNF alpha antibody Mab32 as a template. Species reactivity: Human. Specificity: Recognizes soluble TNF-α. Applications: ELISA, IF, IHC, FUNC. Purity: Protein A affinity purified. Formulation: Liquid. In PBS containing 0.02% Proclin 300.
Catalog No. 50-201-3092