Learn More
Enzo Life Sciences TNF-alpha (soluble) (human), (recombinant) (50 µg)
SDP
Supplier:  Enzo Life Sciences ALX522008C050
Encompass_Preferred
Alternative name: Tumor necrosis factor alpha, TNFSF 2. MW: ~19kDa (SDS-PAGE). Concentration: 1mg/ml after reconstitution. Endotoxin content: E. coli. The extracellular domain of human TNF-α (tumor necrosis factor-α) (aa 85-223) is fused at the N-terminus to a linker peptide (8 aa) and a FLAG-tag. Specificity: Binds to human and mouse TNF-R1 and less efficiently to TNF-R2. In the presence of cross-linking enhancer (Prod. No. ALX-804-034), TNF-α shows a significantly higher affinity for TNF-R2 than for TNF-R1, mimicking the characteristics of membrane-bound TNF-α. Use/Stability: Stable for at least 6 months after receipt when stored at -20°C. UniProt: P01375. Application: ELISA. Handling: Avoid freeze/thaw cycles. After reconstitution, prepare aliquots and store at -20°C. Long Term Storage: -20°C
Catalog No. NC0707578