Learn More
Genscript Corporation Antibody Protein Proteins Proteins 1 MG 1/EA
Supplier:  Genscript Corporation Z033161
Encompass
Tumor Necrosis Factor-Alpha (TNF-Alpha) plays a major role in regulating growth differentiation inflammation viral replication tumorigenesis and autoimmune diseases. TNF alpha-1a is a potent lymphoid factor that exerts cytotoxic effects on a wide range of tumor cells. In addition to inducing hemorrhagic necrosis of tumors studies indicate TNF is involved in tumor igenesis tumor metastasis viral replication septic shock fever inflammation Crohn s disease rheumatoid arthritis and graft-versus-host disease.
Catalog No. 50-252-9665