Learn More
Medchemexpress LLC HY-P7302 50ug Medchemexpress, TNF-alpha, Bovine CAS: Purity:>98%
SDP
Supplier:  Medchemexpress LLC HYP730250UG
Encompass_Preferred
Medchemexpress, HY-P7302 50ug TNF-alpha, Bovine CAS: Recombinant Bovine Tumor Necrosis Factor alpha is a cytokine involved in systemic inflammation and is one of the cytokines that make up the acute phase reaction. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-193-4376