Learn More
Enzo Life Sciences TNF-alpha (human), ELISA kit (1x96wells)
SDP
Supplier:  Enzo Life Sciences ADI900099
Encompass_Preferred
Highly sensitive ELISA for measuring TNF-α, the primary mediator of immune regulation. The TNF-α (human), EIA kit is a colorimetric immunometric enzyme immunoassay kit with results in 4 hours. Absorbance is read at 450 nm. Assay up to 39 samples in duplicate per 96-well plate. The broad dynamic range makes assaying different pathologies easy. Alternative name: Tumor necrosis factor-alpha, TNFSF 2. Sensitivity: 8.43 pg/ml (range 15.63 - 1,000 pg/ml). Use/Stability: Store all components at +4°C, except standard at -20°C. Assay Time: 4 hours. Applications: ELISA, Colorimetric detection . Species reactivity: Human .
Catalog No. 50-200-6841