Learn More
Enzo Life Sciences TNF-alpha (mouse), ELISA kit (1x96wells)
SDP
Supplier:  Enzo Life Sciences ADI900047
Encompass_Preferred
The TNF-α (mouse), EIA kit is a colorimetric immunometric enzyme immunoassay kit with results in 5 hours. Absorbance is read at 450 nm. Alternative name: Tumor necrosis factor-alpha, TNFSF 2. Sensitivity: 3.9 pg/ml (range 31.25 - 2,000 pg/ml). Use/Stability: Store all components at 4°C, except standard at -20°C. UniProt ID: P06804. Assay Time: 5 hours. Applications: ELISA, Colorimetric detection . Species reactivity: Mouse . Content: Microtiter plate, Conjugate, Antibody, Assay buffer 16, Wash buffer concentrate, Standard, TMB Substrate, Stop solution.
Catalog No. 50-200-6831