Learn More
AdipoGen anti-TNF-alpha (mouse), mAb (blocking) (V1q) (preservative free) (500 µg)
SDP
Supplier:  AdipoGen AG20B0081PFC500
Encompass_Preferred
Monoclonal Antibody. Recognizes mouse TNF-alpha. Reacts with both natural and recombinant mouse TNF-alpha. It does not cross-react with human TNF-alpha and TNF-beta (Lymphotoxin-alpha). Application: FACS, FUNC. Clone: V1q. Isotype: Rat IgG. Liquid. In PBS. Tumor necrosis factor (TNF, cachexin or cachectin and formerly known as tumor necrosis factor-α) is a cytokine involved in systemic inflammation and metabolism and is a member of a group of cytokines that stimulate the acute phase reaction. It causes tumor necrosis in vivo and shows also cytolytic activity against tumors. Signal transduction occurs via two types of TNF-receptors, the TNF-receptors 1 and 2. The receptor TNF-R1 is activated by both the membrane-bound and soluble trimeric forms of TNF-α, whereas the receptor TNF-R2 only responds to the membrane-bound form of TNF-α. The monoclonal antibody V1q recognizes endogenous mouse Tumor Necrosis Factor-α.
Catalog No. 50-205-2998