Learn More
Enzo Life Sciences TNF-alpha polyclonal antibody (100µg)
SDP
Supplier:  Enzo Life Sciences ALX210335C100
Encompass_Preferred
Host: Rabbit. Immunogen: Recombinant rat TNF-α (tumor necrosis factor-α). UniProt ID: P16599. Species reactivity: Human, Mouse, Rat. Applications: ELISA, WB, FUNC. Recommended Dilutions: Western Blot (10ng recombinant rat TNF-α by 40-60µg/ml PAb to TNF-α, ALX-210-335) Suggested dilutions/conditions may not be available for all applications. Optimal conditions must be determined individually for each application. Formulation: Liquid. In PBS. Handling: Avoid freeze/thaw cycles. After opening, prepare aliquots and store at -20°C.
Catalog No. 50-200-8113