Learn More
Genscript Corporation Antibody Protein Proteins Proteins 1 MG 1/EA
Supplier:  Genscript Corporation Z033381
Encompass
Tumor Necrosis Factor-Alpha (TNF-alpha) plays a major role in growth regulation differentiation inflammation viral replication tumorigenesis and autoimmune disease. Besides inducing hemorrhagic necrosis of tumors TNF has been found to be involved in tumorigenesis tumor metastasis viral replication septic shock fever inflammation and autoimmune disease including Crohn s disease rheumatoid arthritis and graft-versus-host disease. TNF alpha-1a is a potent lymphoid factor that exerts cytotoxic effects on a wide range of tumor cells and certain other target cells.
Catalog No. 50-252-9658