Learn More
Sino Biological Recombinant Mouse TNF-alpha Protein, Amine-Labeled Biotinylated
Supplier:  Sino Biological 50349MNAEB100
Encompass_Preferred
A DNA sequence encoding the mouse TNF (NP_038721.1) (Leu80-Leu235) was expressed, with an initial Met at the N-terminus. The purified protein was biotinylated in vitro.
Catalog No. 50-211-6214