Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ProSci TNF-alpha Recombinant Protein
SDP

Catalog No. 16838532
Change view
Click to view available options
Quantity:
0.01 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
16-838-532 0.01 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 16-838-532 Supplier ProSci Supplier No. 401150010
Only null left
Add to Cart
Add to Cart

Recombinant Protein

TNF-α is a pleiotrophic pro-inflammatory cytokine secreted by various cells including adipocytes, activated monocytes, macrophages, B cells, T cells and fibroblasts. It belongs to the TNF family of ligands and signals through two receptors, TNFR1 and TNFR2. TNF-α is cytotoxic to a wide variety of tumor cells and is an essential factor in mediating the immune response against bacterial infections. TNF-α also plays a role in the induction of septic shock, auto immune diseases, rheumatoid arthritis, inflammation, and diabetes. Recombinant human TNF-α is a soluble 157 amino acid protein (17.4 kDa) which corresponds to C-terminal extracellular domain of the full length transmembrane protein.

Specifications

Accession Number NP_000585.2
Gene ID (Entrez) 7124
Quantity 0.01 mg
Source E. coli
Common Name tumor necrosis factor
Gene Symbol TNF
Biological Activity The ED50 was determined by the cytolysis of murine L929 cells in the presence of Actinomycin D is ≤ 0.05 ng/mL, corresponding to a specific activity of ≥ 2 x 107 units/mg.
Conjugate Unconjugated
Species Human
Recombinant Recombinant
Sequence VRSSSRTPSD KPVAHVVANP QAEGQLQWLN RRANALLANG VELRDNQLVV PSEGLYLIYS QVLFKGQGCP STHVLLTHTI SRIAVSYQTK VNLLSAIKSP CQRETPEGAE AKPWYEPIYL GGVFQLEKGD RLSAEINRPD YLDFAESGQV YFGIIAL
Content And Storage The lyophilized TNF-alpha recombinant protein is stable for at least 2 years from date of receipt at -20°C. Reconstituted TNF-alpha is stable for at least 3 months when stored in working aliquots with a carrier protein at -20°C. As with any protein, exposing TNF-alpha recombinant protein to repeated freeze / thaw cycles is not recommended. When working with proteins care should be taken to keep recombinant protein (at a cool and stable temperature.)
Purity or Quality Grade >98% by SDS-PAGE gel and HPLC analyses. Endotoxin level is less than 0.1 ng / μg (1 EU/μg).
Synonym DIF, TNFA, TNFSF2, TNF-alpha, Tumor necrosis factor, Cachectin, TNF-a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.