Learn More
Medchemexpress LLC TNF-alpha/TNFSF2 Cy 10ug | 10ug
SDP
Catalog No. 5000187201
Encompass_Preferred
Tumour Necrosis Factor alpha (TNF alpha) is a potent pro-inflammatory cytokine[1] TNF alpha binds to its receptors mainly TNFR1 and TNFR2 and then transmits molecular signals for biological functions such as inflammation and cell death[2] TNF alpha stimulates NF- B pathway via TNFR2 promotes cancer growth invasion and metastasis Anti-TNF- MAb significantly suppresses the tumor development in colitis-associated cancer (CAC) mouse[3] TNF alpha as a proneurogenic factor activates the SAPK/JNK Pathway and can facilitate neuronal replacement and brain repair in response to brain injury[4] TNF-alpha/TNFSF2 Protein Cynomolgus (Biotinylated) is a biotinylated recombinant protein consisting of 157 amino acids (V77-L233) and is produced in E coli
Catalog No. 50-001-87201 Supplier Medchemexpress LLC Supplier No. HYP7754310UG
Tumour Necrosis Factor alpha (TNF alpha) is a potent pro-inflammatory cytokine[1] TNF alpha binds to its receptors mainly TNFR1 and TNFR2 and then transmits molecular signals for biological functions such as inflammation and cell death[2] TNF alpha stimulates NF- B pathway via TNFR2 promotes cancer growth invasion and metastasis Anti-TNF- MAb significantly suppresses the tumor development in colitis-associated cancer (CAC) mouse[3] TNF alpha as a proneurogenic factor activates the SAPK/JNK Pathway and can facilitate neuronal replacement and brain repair in response to brain injury[4] TNF-alpha/TNFSF2 Protein Cynomolgus (Biotinylated) is a biotinylated recombinant protein consisting of 157 amino acids (V77-L233) and is produced in E coli