Learn More
Medchemexpress LLC TNF-alpha/TNFSF2 Cy 2ug
SDP
Catalog No. 5000180690
Encompass_Preferred
TNF-alpha/TNFSF2 protein binds to TNFRSF1A/TNFR1 and TNFRSF1B/TNFBR It induces tumor cell death fever cachexia cell proliferation and cell differentiation It also causes insulin resistance and GKAP42 protein degradation in adipocytes Additionally it plays a role in angiogenesis osteoclastogenesis and IL12 production in dendritic cells TNF-alpha/TNFSF2 Protein Cynomolgus (N-His) is the recombinant cynomolgus-derived TNF-alpha/TNFSF2 protein expressed by E coli with N-6 His labeled tag
Catalog No. 50-001-80690 Supplier Medchemexpress LLC Supplier No. HYP73444A2UG
TNF-alpha/TNFSF2 protein binds to TNFRSF1A/TNFR1 and TNFRSF1B/TNFBR It induces tumor cell death fever cachexia cell proliferation and cell differentiation It also causes insulin resistance and GKAP42 protein degradation in adipocytes Additionally it plays a role in angiogenesis osteoclastogenesis and IL12 production in dendritic cells TNF-alpha/TNFSF2 Protein Cynomolgus (N-His) is the recombinant cynomolgus-derived TNF-alpha/TNFSF2 protein expressed by E coli with N-6 His labeled tag