Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC TNF-alpha/TNFSF2 Hu 50ug
SDP

Catalog No. 5000184844
Encompass_Preferred

Recombinant human TNF-alpha (TNFSF2) supplied as a lyophilized protein corresponding to residues V77-L233 (157 amino acids). The protein is expressed in E. coli, purified to high purity, and provided in small research quantities for in vitro studies of inflammation, apoptosis, and NF-κB signaling. Store lyophilized at -20°C; after reconstitution, store at 4°C short term or -20°C for longer-term storage.

  • Recombinant human TNF-alpha fragment (V77-L233).
  • Expressed in E. coli for high-yield production.
  • Lyophilized powder for long-term storage.
  • Purity greater than 95% as determined by reducing SDS-PAGE.
  • Predicted molecular mass approximately 17.5 kDa.
  • Suitable for in vitro studies of inflammation, apoptosis, and NF-κB signaling.
  • Available in small research quantities, including 50 μg.

Catalog No. 50-001-84844 Supplier Medchemexpress LLC Supplier No. HYP7058A50UG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Recombinant human TNF-alpha (TNFSF2) supplied as a lyophilized protein corresponding to residues V77-L233 (157 amino acids). The protein is expressed in E. coli, purified to high purity, and provided in small research quantities for in vitro studies of inflammation, apoptosis, and NF-κB signaling. Store lyophilized at -20°C; after reconstitution, store at 4°C short term or -20°C for longer-term storage.

  • Recombinant human TNF-alpha fragment (V77-L233).
  • Expressed in E. coli for high-yield production.
  • Lyophilized powder for long-term storage.
  • Purity greater than 95% as determined by reducing SDS-PAGE.
  • Predicted molecular mass approximately 17.5 kDa.
  • Suitable for in vitro studies of inflammation, apoptosis, and NF-κB signaling.
  • Available in small research quantities, including 50 μg.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.